Zirconyl nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
243493-100G 243493-500G 243493-5G
SIAL243493-100GEA 124 EUR
SIAL243493-100G SIAL243493-500G SIAL243493-5G
Zirconyl nitrate hydrate, Sigma-Aldrich®
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III