1-Iodooctane 98% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Octyl iodide

412461000 412465000
ACRO412461000EA 82.5 EUR
ACRO412461000 ACRO412465000
1-Iodooctane 98% stabilised
1-Iodooctane
Formula: C₈H₁₇I
Boiling Pt: 225…226 °C (760 mmHg)
Melting Pt: 46…45 °C
Density: 1.33 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00001105
CAS Number: 629-27-6
EINECS: 211-084-5

Order Now

Specification Test Results

Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4868 to 1.4888 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper
Acidity (CH3COOH) ≤0.002 %
Peroxides ≤0.005 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR