1-Cetylpyridinium chloride monohydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
M-Clarity™ quality level = MQ200
C9002-500G C9002-100G C9002-1KG C9002-25G
SIALC9002-500GEA 203 EUR
SIALC9002-500G SIALC9002-100G SIALC9002-1KG SIALC9002-25G
1-Cetylpyridinium chloride monohydrate, Sigma-Aldrich®
1-Cetylpyridinium chloride monohydrate
Formula: C₂₁H₃₈ClN·1H₂O
MW: 358.01 g/mol
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,I
Merck Index: 13,02041