(R)-(+)-1-Phenylethyl isocyanate, Sigma-Aldrich®

Supplier: Merck

Synonyms: (R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate

M-Clarity™ quality level = MQ200

220574-1G 220574-5G
SIAL220574-1GEA 52.2 EUR
SIAL220574-1G SIAL220574-5G
(R)-(+)-1-Phenylethyl isocyanate, Sigma-Aldrich®
(R)-(+)-1-Phenylethyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 74…76 °C (7 hPa)
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739387
CAS Number: 33375-06-3
EINECS: 251-485-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR