Trichloroacetyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
217328-10G 217328-1G
SIAL217328-10GEA 129 EUR
SIAL217328-10G SIAL217328-1G
Trichloroacetyl isocyanate, Sigma-Aldrich®
Trichloroacetyl isocyanate
Formula: C₃Cl₃NO₂
Boiling Pt: 80…85 °C (27 hPa)
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00002033
CAS Number: 3019-71-4
EINECS: 221-165-7
UN: 2927
ADR: 6.1,II