Trichloroacetic anhydride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

367710-100G 367710-25G
SIAL367710-100GEA 288 EUR
SIAL367710-100G SIAL367710-25G
Trichloroacetic anhydride, Sigma-Aldrich®
Trichloroacetic anhydride
Formula: C₄Cl₆O₃
Boiling Pt: 222…224 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.6908 g/cm³ (20 °C)
MDL Number: MFCD00000793
CAS Number: 4124-31-6
EINECS: 223-925-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR