Formic acid 0.1% in acetonitrile
Supplier: Roth Carl
|
Danger
|
Synonyms:
Aminic Acid, Methanoic acid
CP00.2
CP00.1
ROTHCP00.2EA
267
EUR
ROTHCP00.2
ROTHCP00.1
Formic acid 0.1% in acetonitrile
Formic acid
Formula:
HCO₂H MW: 46.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...