Malononitrile, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
690414-10KG 690414-1KG 690414-50KG
SIAL690414-10KGEA 1610 EUR
SIAL690414-10KG SIAL690414-1KG SIAL690414-50KG
Malononitrile, Sigma-Aldrich®
Malononitrile
Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.191 g/cm³ (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 13,05733