Malononitrile, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
M1407-100G-A M1407-500G-A M1407-5G-A
SIALM1407-100G-AEA 44 EUR
SIALM1407-100G-A SIALM1407-500G-A SIALM1407-5G-A
Malononitrile, Sigma-Aldrich®
Malononitrile
Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.191 g/cm³ (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 13,05733