tert-Butyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
tert-Butyl Acetoacetate
M-Clarity™ quality level = MQ100
537594-100ML
537594-1L
537594-500ML
537594-4L
SIAL537594-100MLEA
35.9
EUR
SIAL537594-100ML
SIAL537594-1L
SIAL537594-500ML
SIAL537594-4L
tert-Butyl acetoacetate, Sigma-Aldrich®
tert-Butyl acetoacetate
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol Boiling Pt: 190 °C (1013 hPa) Melting Pt: –38 °C Density: 0.9756 g/cm³ (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00008811 CAS Number: 1694-31-1 EINECS: 216-904-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...