Methyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Methyl 3-oxobutanoate
M-Clarity™ quality level = MQ200
537365-100G
537365-1KG
SIAL537365-100GEA
16
EUR
SIAL537365-100G
SIAL537365-1KG
Methyl acetoacetate, Sigma-Aldrich®
Methyl acetoacetate
Formula:
C₅H₈O₃ MW: 116.12 g/mol Boiling Pt: 170 °C (1013 hPa) Melting Pt: –39 °C Density: 1.076 g/cm³ (20 °C) Flash Pt: 67 °C |
MDL Number:
MFCD00008784 CAS Number: 105-45-3 EINECS: 203-299-8 Merck Index: 12,06090 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...