Tributyrin (glycerol tributyrate), Sigma-Aldrich®
Supplier: Merck
Synonyms:
Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate
M-Clarity™ quality level = MQ200
91010-100ML
91010-25ML
SIAL91010-100MLEA
130
EUR
SIAL91010-100ML
SIAL91010-25ML
Tributyrin (glycerol tributyrate), Sigma-Aldrich®
Tributyrin (Glycerol tributyrate)
Formula:
C₁₅H₂₆O₆ MW: 302.37 g/mol Boiling Pt: 305 °C (1013 hPa) Melting Pt: –70 °C Density: 1.035 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009392 CAS Number: 60-01-5 EINECS: 200-451-5 Merck Index: 12,09750 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...