Tributyrin (glycerol tributyrate), Sigma-Aldrich®

Supplier: Merck

Synonyms: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate

T8626-100ML T8626-25ML
SIALT8626-100MLEA 181 EUR
SIALT8626-100ML SIALT8626-25ML
Tributyrin (glycerol tributyrate), Sigma-Aldrich®
Tributyrin (Glycerol tributyrate)
Formula: C₁₅H₂₆O₆
MW: 302.37 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: –70 °C
Density: 1.035 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009392
CAS Number: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR