Triacetin (glycerol triacetate), Sigma-Aldrich®
Supplier: Merck
Synonyms:
Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
MQ100
525073-1L
525073-20L
525073-4L
SIAL525073-1LEA
83.1
EUR
SIAL525073-1L
SIAL525073-20L
SIAL525073-4L
Triacetin (glycerol triacetate), Sigma-Aldrich®
Triacetin (Glycerol triacetate)
Formula:
C₉H₁₄O₆ MW: 218.21 g/mol Boiling Pt: 258 Melting Pt: 4 Density: 1.1596 g/cm³ (20 °C) Flash Pt: 138 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008716 CAS Number: 102-76-1 EINECS: 203-051-9 Merck Index: 13,09664 |