Triacetin (glycerol triacetate), Sigma-Aldrich®
Supplier: Merck
Synonyms:
Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
MQ200
90240-1L
90240-250ML
SIAL90240-1LEA
96.5
EUR
SIAL90240-1L
SIAL90240-250ML
Triacetin (glycerol triacetate), Sigma-Aldrich®
Triacetin (Glycerol triacetate)
Formula:
C₉H₁₄O₆ MW: 218.21 g/mol Boiling Pt: 258 Melting Pt: 4 Density: 1.1596 g/cm³ (20 °C) Flash Pt: 138 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008716 CAS Number: 102-76-1 EINECS: 203-051-9 Merck Index: 13,09664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...