Triacetin (glycerol triacetate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
M-Clarity™ quality level = MQ300
W200700-10KG-K W200700-1KG-K W200700-25KG-K
SIALW200700-10KG-KEA 202 EUR
SIALW200700-10KG-K SIALW200700-1KG-K SIALW200700-25KG-K
Triacetin (glycerol triacetate), Sigma-Aldrich®
Triacetin (Glycerol triacetate)
Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.1596 g/cm³ (20 °C)
Flash Pt: 138 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664