Allyl enantate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Heptylic acid allyl ester, Enanthic acid allyl ester, Allyl Heptanoate, Allyl heptanoate, Heptanoic acid allyl ester
M-Clarity™ quality level = MQ300
W203106-1KG-K W203106-4KG-K W203106-9KG-K W203106-SAMPLE
SIALW203106-1KG-KEA 95.2 EUR
SIALW203106-1KG-K SIALW203106-4KG-K SIALW203106-9KG-K SIALW203106-SAMPLE
Allyl enantate, Sigma-Aldrich®
Allyl enantate
Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 210 °C (1013 hPa)
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00038341
CAS Number: 142-19-8
EINECS: 205-527-1
UN: 2810
ADR: 6.1,III