Hexyl butyrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W256803-1KG-K W256803-20KG-K W256803-9KG-K
SIALW256803-1KG-KEA 86.9 EUR
SIALW256803-1KG-K SIALW256803-20KG-K SIALW256803-9KG-K
Hexyl butyrate, Sigma-Aldrich®
Hexyl butyrate
Formula: C₁₀H₂₀O₂
Boiling Pt: 205 °C (1013 hPa)
Density: 0.865 g/cm³ (20 °C)
MDL Number: MFCD00048884
CAS Number: 2639-63-6
EINECS: 220-136-6