2-Methylbutyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetic acid 2-Methylbutyl ester
M-Clarity™ quality level = MQ300
W364401-1KG-K W364401-4KG-K W364401-9KG-K
SIALW364401-1KG-KEA 88.5 EUR
SIALW364401-1KG-K SIALW364401-4KG-K SIALW364401-9KG-K
2-Methylbutyl acetate, Sigma-Aldrich®
2-Methylbutyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 140…142 °C (1013 hPa)
Density: 0.876 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040494
CAS Number: 624-41-9
EINECS: 210-843-8
UN: 1104
ADR: 3,III