Ethyl enantate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Heptylic acid ethyl ester, Enanthic acid ethyl ester, Heptanoic acid ethyl ester, Ethyl heptanoate
M-Clarity™ quality level = MQ300
W243728-100G-K W243728-1KG-K
SIALW243728-100G-KEA 130 EUR
SIALW243728-100G-K SIALW243728-1KG-K
Ethyl enantate, Sigma-Aldrich®
Ethyl enantate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 372 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00009538
CAS Number: 106-30-9
EINECS: 203-382-9
Merck Index: 13,03867