2-Butoxyethyl acetate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Butylglycol acetate
M-Clarity™ quality level = MQ200
307289-1L 307289-25ML
SIAL307289-1LEA 64.7 EUR
SIAL307289-1L SIAL307289-25ML
2-Butoxyethyl acetate, Sigma-Aldrich®
2-Butoxyethyl acetate
Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 192
Melting Pt: –2 °C
Density: 0.9416 g/cm³ (20 °C)
Flash Pt: 82
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3