Lithium acetate dihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
L4158-100G L4158-1KG L4158-250G
SIALL4158-100GEA 66.9 EUR
SIALL4158-100G SIALL4158-1KG SIALL4158-250G
Lithium acetate dihydrate, Sigma-Aldrich®
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543