Sodium iodoacetate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Sodium 2-iodoacetate, Iodoacetic Acid, Sodium Salt
M-Clarity™ quality level = MQ200
I9148-25G
I9148-5G
SIALI9148-25GEA
662
EUR
SIALI9148-25G
SIALI9148-5G
Sodium iodoacetate, Sigma-Aldrich®
Sodium iodoacetate
Formula:
C₂H₂INaO₂ Melting Pt: 208…210 °C Flash Pt: 208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002686 CAS Number: 305-53-3 EINECS: 206-165-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...