Citric acid
Supplier: Hach
Warning
HACH1454899EA 52.4 EUR
HACH1454899
Citric acid
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MW: 192,13 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1,665 g/cm³ (20 °C)
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350
https://si.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg