Diethylenetriaminepentaacetic acid
Supplier: Molekula
MOLE69522692-1KGEA
210
EUR
MOLE69522692-1KG
Diethylenetriaminepentaacetic acid
Diethylenetriaminepentaacetic acid
Formula:
C₁₄H₂₃N₃O₁₀ MW: 393,35 g/mol Melting Pt: 220…222 °C Density: 1,558 g/cm³ (20 °C) |
MDL Number:
MFCD00004289 UN: 3077 ADR: 9,III Merck Index: 12,07266 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...