Diethylenetriaminepentaacetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
D6518-100G D6518-10G D6518-50G D6518-5G
SIALD6518-100GEA 789 EUR
SIALD6518-100G SIALD6518-10G SIALD6518-50G SIALD6518-5G
Diethylenetriaminepentaacetic acid, Sigma-Aldrich®
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
Melting Pt: 220…222 °C
Density: 1.558 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9,III
Merck Index: 13,07202