Ethyl iodoacetate, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

242934-5G 242934-25G
SIAL242934-5GEA 47.7 EUR
SIAL242934-5G SIAL242934-25G
Ethyl iodoacetate, Sigma-Aldrich®
Ethyl iodoacetate
Formula: C₄H₇IO₂
MW: 214 g/mol
Boiling Pt: 179
Density: 1.81 g/cm³ (20 °C)
MDL Number: MFCD00001081
CAS Number: 623-48-3
EINECS: 210-796-3
UN: 2927
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR