2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Supplier: BIOMOL RESEARCH LABORATORIES
Synonyms: BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid
50002-1 50002-2
BMOL50002-1EA 0 EUR
BMOL50002-1 BMOL50002-2
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formula: C₆H₁₅NO₅S
MW: 213.25 g/mol
Boiling Pt: 405
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5