Diethylenetriamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine
M-Clarity™ quality level = MQ200
D93856-1L D93856-2.5L D93856-5ML D93856-18L
SIALD93856-1LEA 71.6 EUR
SIALD93856-1L SIALD93856-2.5L SIALD93856-5ML SIALD93856-18L
Diethylenetriamine, Sigma-Aldrich®
Diethylenetriamine
Formula: C₄H₁₃N₃
MW: 103.17 g/mol
Boiling Pt: 207
Melting Pt: –39 °C
Density: 0.9523 g/cm³ (20 °C)
Flash Pt: 97
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8 II