Acetone oxime, Sigma-Aldrich®
Supplier: Merck
Synonyms: Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime
M-Clarity™ quality level = MQ200
00750-10G 00750-250G
SIAL00750-10GEA 29.1 EUR
SIAL00750-10G SIAL00750-250G
Acetone oxime, Sigma-Aldrich®
Acetone oxime
Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: 61 °C
Density: 1.045 g/cm³ (20 °C)
MDL Number: MFCD00002118
CAS Number: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,00077