Ethylene glycol monomethyl ether (2-methoxyethanol), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethylene glycol monomethyl ether
M-Clarity™ quality level = MQ100
284467-100ML 284467-1L
SIAL284467-100MLEA 87.7 EUR
SIAL284467-100ML SIAL284467-1L
Ethylene glycol monomethyl ether (2-methoxyethanol), Sigma-Aldrich®
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118