Acetal, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,1-Diethoxyethane, Acetaldehyde diethyl acetal
M-Clarity™ quality level = MQ300
W200220-100G-K
W200220-1KG-K
W200220-4KG-K
SIALW200220-100G-KEA
90.1
EUR
SIALW200220-100G-K
SIALW200220-1KG-K
SIALW200220-4KG-K
Acetal, Sigma-Aldrich®
Acetal
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 102.7 °C (1013 hPa) Melting Pt: –100 °C Density: 0.83 g/cm³ (20 °C) Flash Pt: –21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009243 CAS Number: 105-57-7 EINECS: 203-310-6 UN: 1088 ADR: 3,II Merck Index: 13,00040 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...