Itaconic anhydride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione
M-Clarity™ quality level = MQ200
259926-100G
259926-25G
SIAL259926-100GEA
158
EUR
SIAL259926-100G
SIAL259926-25G
Itaconic anhydride, Sigma-Aldrich®
Itaconic anhydride
Formula:
C₅H₄O₃ MW: 112.08 g/mol Boiling Pt: 239 °C (1013 hPa) Melting Pt: 64…65 °C |
MDL Number:
MFCD00005530 CAS Number: 2170-03-8 EINECS: 218-518-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...