Ethyl 2-bromoisobutyrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester
M-Clarity™ quality level = MQ200
E14403-100G E14403-500G E14403-5G
SIALE14403-100GEA 53.7 EUR
SIALE14403-100G SIALE14403-500G SIALE14403-5G
Ethyl 2-bromoisobutyrate, Sigma-Aldrich®
Ethyl 2-bromoisobutyrate
Formula: C₆H₁₁BrO₂
Boiling Pt: 65 °C (1013 hPa)
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00000123
CAS Number: 600-00-0
EINECS: 209-980-6