Pinacol 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
418001000
418002500
418000010
ACRO418001000EA
130
EUR
ACRO418001000
ACRO418002500
ACRO418000010
Pinacol 99%
Pinacol
Formula:
C₆H₁₄O₂ Boiling Pt: 174 °C Melting Pt: 40…43 °C Density: 0,967 g/cm³ (20 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004462 CAS Number: 76-09-5 EINECS: 200-933-5 Merck Index: 13,07521 |
Specification Test Results
Appearance | White Low melting crystalline solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Titration with NaOH | ≥97.5 % |