2,2,4,4,6,6-Hexamethylheptane

Supplier: Thermo Scientific
Danger

RJC01249.250 RJC01249.1
MAYBRJC01249.250EA 0 EUR
MAYBRJC01249.250 MAYBRJC01249.1
2,2,4,4,6,6-Hexamethylheptane
2,2,4,4,6,6-Hexamethylheptane
Formula: C₁₃H₂₈
MW: 184.37 g/mol
Storage Temperature: Ambient
CAS Number: 34701-49-0
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR