Phenylacetaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Phenyl acetic aldehyde

M-Clarity™ quality level = MQ100

107395-100ML 107395-25ML 107395-500ML
SIAL107395-100MLEA 63.1 EUR
SIAL107395-100ML SIAL107395-25ML SIAL107395-500ML
Phenylacetaldehyde, Sigma-Aldrich®
Phenylacetaldehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 195
Melting Pt: 33…34 °C
Density: 1.026…1.03 g/cm³ (20 °C)
MDL Number: MFCD00006993
CAS Number: 122-78-1
EINECS: 204-574-5
Merck Index: 12,07419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR