Phenylacetaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Phenyl acetic aldehyde
MQ300
W287407-10KG-K W287407-1KG-K W287407-5KG-K
SIALW287407-10KG-KEA 1200 EUR
SIALW287407-10KG-K SIALW287407-1KG-K SIALW287407-5KG-K
Phenylacetaldehyde, Sigma-Aldrich®
Phenylacetaldehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 195
Melting Pt: 33…34 °C
Density: 1.026…1.03 g/cm³ (20 °C)
MDL Number: MFCD00006993
CAS Number: 122-78-1
EINECS: 204-574-5
Merck Index: 12,07419