2-Phenylethyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate
M-Clarity™ quality level = MQ300
W285714-100G-K W285714-1KG-K W285714-25G-K
SIALW285714-100G-KEA 387 EUR
SIALW285714-100G-K SIALW285714-1KG-K SIALW285714-25G-K
2-Phenylethyl acetate, Sigma-Aldrich®
2-Phenylethyl acetate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.0286 g/cm³ (20 °C)
MDL Number: MFCD00008720
CAS Number: 103-45-7
EINECS: 203-113-5