α-Linolenic acid ethyl ester, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, 9Z,12Z,15Z-Octadecatrienoic acid ethyl ester, Ethyl linolenate, (9Z,12Z,15Z)-Ethyl octadeca-9,12,15-trienoate, Linolenic acid ethyl ester, Ethyl α-linolenate, cis,cis,cis-9,12,15-Octadecatrienoic acid ethyl ester, Ethyl cis,cis,cis-9,12,15-octadecatrienoate
M-Clarity™ quality level = MQ100
L2501-1G L2501-500MG L2501-5G
SIALL2501-1GEA 216 EUR
SIALL2501-1G SIALL2501-500MG SIALL2501-5G
α-Linolenic acid ethyl ester, Sigma-Aldrich®
α-Linolenic acid ethyl ester
Formula: C₂₀H₃₄O₂
MW: 306.49 g/mol
Boiling Pt: 218 °C (1013 hPa)
Density: 0.8919 g/cm³ (20 °C)
MDL Number: MFCD07371452
CAS Number: 1191-41-9
EINECS: 214-734-6