α-Linolenic acid methyl ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methyl cis,cis,cis-9,12,15-octadecatrienoate, Methyl linolenate, 9Z,12Z,15Z-Octadecatrienoic acid methyl ester, Methyl Linolenate, Linolenic acid methyl ester, cis,cis,cis-9,12,15-Octadecatrienoic acid methyl ester, Methyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, Methyl α-linolenate

M-Clarity™ quality level = MQ200

L2626-100MG L2626-500MG L2626-5G L2626-1G
SIALL2626-100MGEA 52.4 EUR
SIALL2626-100MG SIALL2626-500MG SIALL2626-5G SIALL2626-1G
α-Linolenic acid methyl ester, Sigma-Aldrich®
α-Linolenic acid methyl ester
Formula: C₁₉H₃₂O₂
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.8982 g/cm³ (20 °C)
MDL Number: MFCD00135851
CAS Number: 301-00-8
EINECS: 206-102-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR