γ-Linolenic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid
M-Clarity™ quality level = MQ100
L2378-100MG
L2378-10MG
L2378-1G
L2378-500MG
SIALL2378-100MGEA
151
EUR
SIALL2378-100MG
SIALL2378-10MG
SIALL2378-1G
SIALL2378-500MG
γ-Linolenic acid, Sigma-Aldrich®
γ-Linolenic acid
Formula:
C₁₈H₃₀O₂ MW: 278.44 g/mol |
MDL Number:
MFCD00065718 CAS Number: 506-26-3 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...