α-Linolenic acid
Supplier: Cayman Chemical
Synonyms: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
90210-100 90210-250 90210-50
CAYM90210-100EA 51 EUR
CAYM90210-100 CAYM90210-250 CAYM90210-50
α-Linolenic acid
α-Linolenic acid
Formula: C₁₈H₃₀O₂
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: –9.5 °C
Density: 0.9141 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065720
CAS Number: 463-40-1
EINECS: 207-334-8
Merck Index: 12,05530