α-Linolenic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
M-Clarity™ quality level = MQ200
L2376-10G
L2376-500MG
L2376-5G
SIALL2376-10GEA
1730
EUR
SIALL2376-10G
SIALL2376-500MG
SIALL2376-5G
α-Linolenic acid, Sigma-Aldrich®
α-Linolenic acid
Formula:
C₁₈H₃₀O₂ MW: 278.44 g/mol Boiling Pt: 443 °C (1013 hPa) Melting Pt: –9.5 °C Density: 0.9141 g/cm³ (18 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00065720 CAS Number: 463-40-1 EINECS: 207-334-8 Merck Index: 12,05530 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...