3-Chloro-1-butene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C29002-100ML
C29002-5ML
SIALC29002-100MLEA
348
EUR
SIALC29002-100ML
SIALC29002-5ML
3-Chloro-1-butene, Sigma-Aldrich®
3-Chloro-1-butene
Formula:
C₄H₇Cl MW: 90.55 g/mol Boiling Pt: 64…65 °C (1013 hPa) Melting Pt: –50 °C Density: 0.8978 g/cm³ (20 °C) |
MDL Number:
MFCD00000869 CAS Number: 563-52-0 EINECS: 209-252-8 UN: 1993 ADR: 3 II Merck Index: 13,02150 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...