3-Buten-2-one, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Vinyl methyl ketone, MVK, Methyl vinyl ketone, Butenone

M-Clarity™ quality level = MQ200

M87509-100ML M87509-500ML
SIALM87509-100MLEA 106 EUR
SIALM87509-100ML SIALM87509-500ML
3-Buten-2-one, Sigma-Aldrich®
3-Buten-2-one
Formula: C₄H₆O
Boiling Pt: 81.4 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.8636 g/cm³ (20 °C)
Flash Pt: –7 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00008777
CAS Number: 78-94-4
EINECS: 201-160-6
UN: 1251
ADR: 6.1,I
Merck Index: 12,06214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c0b910689fbbbc834d57744b46744430d66145e&vid=accba25777bad1758e027949bef9960ba106c529&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR