Dimethyl maleate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester
M-Clarity™ quality level = MQ200
238198-100G 238198-1KG
SIAL238198-100GEA 31.6 EUR
SIAL238198-100G SIAL238198-1KG
Dimethyl maleate, Sigma-Aldrich®
Dimethyl maleate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 205
Melting Pt: –17.5 °C
Density: 1.153 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5