β-(N-Morpholino)ethanesulphonic acid (MES)
Supplier: BIOMOL RESEARCH LABORATORIES
Synonyms: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
06010-1 06010-2
BMOL06010-1EA 0 EUR
BMOL06010-1 BMOL06010-2
β-(N-Morpholino)ethanesulphonic acid (MES)
β-(N-Morpholino)ethanesulphonic acid (MES)
Formula: C₆H₁₃NO₄S
Melting Pt: 316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006181
CAS Number: 4432-31-9
EINECS: 224-632-3
Merck Index: 13,05929