γ-(N-Morpholino)propanesulphonic acid (MOPS)
Supplier: PanReac AppliChem
A2947.0500 A2947.1000
A2947.0500EA 374 EUR
A2947.0500 A2947.1000
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288