γ-(N-Morpholino)propanesulphonic acid (MOPS)
Supplier: BIOMOL RESEARCH LABORATORIES
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
06170-1 06170-2 06170-3
BMOL06170-1EA 0 EUR
BMOL06170-1 BMOL06170-2 BMOL06170-3
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288